2-Bromo-5-pyridineboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 2-Bromopyridine-5-boronic acid pinacol ester, 2-Bromo-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

A120112-10G A120112-250MG A120112-25G A120112-1G A120112-5G
103799-888EA 15 USD
103799-888 76797-294 76797-296 76797-298 77297-738 77297-740
2-Bromo-5-pyridineboronic acid pinacol ester 97%
2-Bromo-5-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₅BBrNO₂
MW: 283.96 g/mol
Boiling Pt: 345.1 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD03412794
CAS Number: 214360-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR