2-Bromo-4-nitrotoluene 98%

Supplier: Ambeed
Warning

A150316-100G A150316-10G A150316-500G A150316-5G A150316-25G
103842-278EA 9 USD
103842-278 77137-734 77137-736 77137-738 77299-792 77317-788
2-Bromo-4-nitrotoluene 98%
2-Bromo-4-nitrotoluene

Precaución: for Research Use Only

Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Melting Pt: 76…77 °C
Flash Pt: 123.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00007195
CAS Number: 7745-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR