2-Bromo-4-nitropyridine 97%

Supplier: Ambeed
Warning

A258535-100G A258535-10G A258535-1G A258535-25G A258535-5G
103812-380EA 11 USD
103812-380 103852-258 76988-128 76988-130 77091-786 77091-788 77091-790
2-Bromo-4-nitropyridine 97%
2-Bromo-4-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₃BrN₂O₂
Boiling Pt: 286 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD03701383
CAS Number: 6945-67-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR