2-Bromo-4-nitrobenzoic acid 98%

Supplier: Ambeed
Warning

A126191-100G A126191-10G A126191-1G A126191-25G A126191-5G
103800-772EA 9 USD
103800-772 77258-286 77258-288 77312-954 77312-956 77312-958
2-Bromo-4-nitrobenzoic acid 98%
2-Bromo-4-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄BrNO₄
MW: 246.02 g/mol
Melting Pt: 169 °C
Flash Pt: 184.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00234252
CAS Number: 16426-64-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR