2-Bromo-4-nitrobenzaldehyde 98%

Supplier: Ambeed
Warning

A105203-100MG A105203-250MG A105203-1G A105203-5G
103796-854EA 54 USD
103796-854 103822-582 77257-410 77257-412 77309-456 77309-458
2-Bromo-4-nitrobenzaldehyde 98%
2-Bromo-4-nitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄BrNO₃
Boiling Pt: 553.5 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD08753661
CAS Number: 5274-71-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR