2-Bromo-4-nitro-1-(trifluoromethyl)benzene 97%

Supplier: Ambeed
Warning

A125988-10G A125988-250MG A125988-1G A125988-5G
103800-748EA 34 USD
103800-748 76971-882 76971-884 77312-912 77312-914
2-Bromo-4-nitro-1-(trifluoromethyl)benzene 97%
2-Bromo-4-nitro-1-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₇H₃BrF₃NO₂
MW: 270 g/mol
Boiling Pt: 257.5±40.0 °C (760 mm Hg)
Flash Pt: 109.6 °C
Storage Temperature: Ambient
MDL Number: MFCD16606317
CAS Number: 875238-74-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR