2-Bromo-4-methylthiazole-5-carboxylic acid 95%
Supplier: Ambeed
|
Warning
|
A147532-10G
A147532-25G
A147532-1G
A147532-5G
103803-794EA
7
USD
103803-794
103841-898
77259-100
77259-102
77299-590
77317-274
2-Bromo-4-methylthiazole-5-carboxylic acid 95%
2-Bromo-4-methylthiazole-5-carboxylic acid
Precaución: for Research Use Only
Formula:
C₅H₄BrNO₂S Boiling Pt: 349.5 °C (760 mm Hg) Melting Pt: 158 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09026784 CAS Number: 40003-41-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...