2-Bromo-4-methylthiazole 97%

Supplier: Ambeed
Danger

A172846-1G A172846-5G A172846-10G A172846-25G
103806-756EA 15 USD
103806-756 103845-048 77082-262 77082-264 77163-622 77301-224
2-Bromo-4-methylthiazole 97%
2-Bromo-4-methylthiazole

Precaución: for Research Use Only

Formula: C₄H₄BrNS
Boiling Pt: 195.1 °C (760 mm Hg)
Flash Pt: 71.8 °C
Storage Temperature: Ambient
MDL Number: MFCD06660130
CAS Number: 7238-61-1
UN: 1760
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR