2-Bromo-4-methylpyrimidine 97%

Supplier: Ambeed
Warning

A214187-100G A214187-10G A214187-250MG A214187-1G A214187-25G A214187-5G
103818-376EA 19 USD
103818-376 103849-084 77013-032 77013-034 77013-036 77087-592 77087-594 77087-596
2-Bromo-4-methylpyrimidine 97%
2-Bromo-4-methylpyrimidine

Precaución: for Research Use Only

Formula: C₅H₅ClN₂
Boiling Pt: 265 °C (760 mm Hg)
Melting Pt: 45…50 °C
Flash Pt: 114 °C
Storage Temperature: Ambient
MDL Number: MFCD04037883
CAS Number: 130645-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR