2-Bromo-4-methylpyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-4-picoline

A298751-5G A298751-100G A298751-10G A298751-1G A298751-500G
103835-342EA 6 USD
103835-342 76838-044 77114-420 77114-422 77114-424 77114-426
2-Bromo-4-methylpyridine 98%
2-Bromo-4-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₆BrN
MW: 172.02 g/mol
Boiling Pt: 219.8 °C (760 mm Hg)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00082590
CAS Number: 4926-28-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR