2-Bromo-p-toluonitrile 98%

Supplier: Ambeed
Warning

A185680-25G A185680-5G A185680-100G A185680-10G
103846-444EA 6 USD
103846-444 77084-074 77084-076 77260-372 77260-374
2-Bromo-p-toluonitrile 98%
2-Bromo-p-toluonitrile

Precaución: for Research Use Only

Formula: C₈H₆BrN
MW: 196.05 g/mol
Melting Pt: 50…55 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00158968
CAS Number: 42872-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR