2-Bromo-p-toluic acid 97%

Supplier: Ambeed
Warning

A193936-100G A193936-10G A193936-500G A193936-1G A193936-25G A193936-5G
103816-598EA 6 USD
103816-598 103847-206 77260-580 77260-582 77260-584 77302-372 77302-374 77302-376
2-Bromo-p-toluic acid 97%
2-Bromo-p-toluic acid

Precaución: for Research Use Only

Formula: C₈H₇BrO₂
Melting Pt: 143…147 °C
Flash Pt: 147.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00045797
CAS Number: 7697-27-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR