2-Bromo-4-methylbenzaldehyde 97%

Supplier: Ambeed
Warning

A147827-100G A147827-10G A147827-500G A147827-1G A147827-25G A147827-5G
103803-840EA 6 USD
103803-840 103841-948 77235-272 77235-274 77235-276 77317-360 77317-362 77317-364
2-Bromo-4-methylbenzaldehyde 97%
2-Bromo-4-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇BrO
MW: 199.05
Melting Pt: 30…35 °C
Storage Temperature: Ambient
MDL Number: MFCD04039890
CAS Number: 824-54-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR