2-Bromo-4-methyl-6-nitroaniline 98%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-6-nitro-p-toluidine, 4-Amino-3-bromo-5-nitrotoluene

A160600-100G A160600-10G A160600-250MG A160600-500G A160600-5G A160600-1G A160600-25G
103805-396EA 9 USD
103805-396 103843-628 77138-126 77138-128 77138-130 77138-132 77300-476 77319-576 77319-578
2-Bromo-4-methyl-6-nitroaniline 98%
2-Bromo-4-methyl-6-nitroaniline

Precaución: for Research Use Only

Formula: C₇H₇BrN₂O₂
Boiling Pt: 329.1 °C (760 mm Hg)
Melting Pt: 64…66 °C
Flash Pt: 152.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00209452
CAS Number: 827-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR