2-Bromo-5-nitro-4-picoline 98%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-4-methyl-5-nitropyridine

A290869-25G A290869-5G A290869-100G A290869-10G A290869-500G
103835-022EA 12 USD
103835-022 76837-608 76837-610 77099-318 77099-320 77099-322
2-Bromo-5-nitro-4-picoline 98%
2-Bromo-5-nitro-4-picoline

Precaución: for Research Use Only

Formula: C₆H₅BrN₂O₂
MW: 217.02
Boiling Pt: 294.2 °C (760 mm Hg)
Melting Pt: 64…66 °C
Flash Pt: 131.8 °C
Storage Temperature: Ambient
MDL Number: MFCD03095066
CAS Number: 23056-47-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR