2-Bromo-4-methylimidazole 97%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-4-methyl-1H-imidazole

A133274-250MG A133274-1G A133274-25G
103801-854EA 21 USD
103801-854 77258-552 77314-488 77314-490
2-Bromo-4-methylimidazole 97%
2-Bromo-4-methylimidazole

Precaución: for Research Use Only

Formula: C₄H₅BrN₂
MW: 161 g/mol
Boiling Pt: 280.7 °C (760 mm Hg)
Flash Pt: 123.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06659904
CAS Number: 23328-88-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR