2-Bromo-4-methoxypyridine 98%

Supplier: Ambeed
Warning

A130368-10G A130368-250MG A130368-1G A130368-25G A130368-5G
103801-412EA 22 USD
103801-412 76870-688 76870-690 77313-842 77313-844 77313-846
2-Bromo-4-methoxypyridine 98%
2-Bromo-4-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₆BrNO
Boiling Pt: 231.1 °C (760 mm Hg)
Flash Pt: 93.5 °C
Storage Temperature: Ambient
MDL Number: MFCD06254613
CAS Number: 89488-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR