2-Bromo-4-methoxyphenol 97%

Supplier: Ambeed
Warning

Synonyms: 3-Brom-4-hydroxyanisole

A235974-25G A235974-5G A235974-100G A235974-10G A235974-1G
103811-058EA 9 USD
103811-058 103850-828 77089-860 77089-862 77140-166 77140-168 77304-028
2-Bromo-4-methoxyphenol 97%
2-Bromo-4-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₇BrO₂
Boiling Pt: 245.5 °C (760 mm Hg)
Melting Pt: 47 °C
Flash Pt: 102.2 °C
Storage Temperature: Ambient
MDL Number: MFCD02113712
CAS Number: 17332-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR