2-Bromo-4-methoxybenzonitrile 95%
Supplier: Ambeed
|
Warning
|
A213836-10G
A213836-1G
A213836-5G
A213836-250MG
103818-342EA
6
USD
103818-342
103849-054
77030-586
77087-558
77087-560
77671-906
2-Bromo-4-methoxybenzonitrile 95%
2-Bromo-4-methoxybenzonitrile
Precaución: for Research Use Only
Formula:
C₈H₆BrNO MW: 212.0433 g/mol Boiling Pt: 336.4 °C (760 mm Hg) Melting Pt: 212…214 °C Storage Temperature: Ambient |
MDL Number:
MFCD09056764 CAS Number: 140860-51-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...