2-Bromo-p-anisaldehyde 98%

Supplier: Ambeed
Warning

A153271-100G A153271-10G A153271-500G A153271-1G A153271-25G A153271-5G
103804-484EA 9 USD
103804-484 103842-656 76783-464 76783-466 76783-468 77318-304 77318-306 77318-308
2-Bromo-p-anisaldehyde 98%
2-Bromo-p-anisaldehyde

Precaución: for Research Use Only

Formula: C₈H₇BrO₂
Boiling Pt: 283.9 °C (760 mm Hg)
Melting Pt: 75…77 °C
Flash Pt: 126 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09788636
CAS Number: 43192-31-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR