2-Bromo-p-anisidine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-4-methoxyaniline

A130974-100G A130974-10G A130974-1G A130974-5G
103801-506EA 9 USD
103801-506 77258-446 77258-448 77298-440 77313-984
2-Bromo-p-anisidine 98%
2-Bromo-p-anisidine

Precaución: for Research Use Only

Formula: C₇H₈BrNO
MW: 202.0485 g/mol
Boiling Pt: 265.1 °C (760 mm Hg)
Flash Pt: 114.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09029057
CAS Number: 32338-02-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR