2-Bromo-4-methoxy-5-methylpyridine 97%

Supplier: Ambeed
Warning

A903725-1G A903725-5G A903725-100MG A903725-250MG
103841-356EA 487 USD
103841-356 76845-624 76845-626 77147-126 77147-128
2-Bromo-4-methoxy-5-methylpyridine 97%
2-Bromo-4-methoxy-5-methylpyridine

Precaución: for Research Use Only

Formula: C₇H₈BrNO
Storage Temperature: Ambient
MDL Number: MFCD16610108
CAS Number: 1227574-33-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR