2-Bromo-4-iodopyridine 98%
Supplier: Ambeed
|
Warning
|
A239360-100G
A239360-10G
A239360-250MG
A239360-1G
A239360-25G
A239360-5G
103811-282EA
14
USD
103811-282
103851-080
76987-590
76987-592
76987-594
77090-222
77090-224
77090-226
2-Bromo-4-iodopyridine 98%
Precaución: for Research Use Only
Formula:
C₅H₃BrIN Melting Pt: 61 °C Flash Pt: 127.2 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03085768 CAS Number: 100523-96-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...