2-Bromo-4-iodobenzoic acid 98%

Supplier: Ambeed
Warning

A133979-10G A133979-1G A133979-5G
103801-944EA 6 USD
103801-944 76811-378 77314-642 77314-644
2-Bromo-4-iodobenzoic acid 98%
2-Bromo-4-iodobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄BrIO₂
MW: 326.914 g/mol
Melting Pt: 183…187 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11520409
CAS Number: 28547-29-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR