2-Bromo-4-iodoaniline 98%

Supplier: Ambeed
Warning

A189884-1G A189884-5G A189884-100G A189884-10G A189884-250MG A189884-25G
103816-228EA 6 USD
103816-228 103846-824 77084-594 77084-596 77260-478 77260-480 77260-482 77260-484
2-Bromo-4-iodoaniline 98%
2-Bromo-4-iodoaniline

Precaución: for Research Use Only

Formula: C₆H₅BrIN
MW: 297.9200 g/mol
Boiling Pt: 307.6 °C (760 mm Hg)
Melting Pt: 75…76 °C
Flash Pt: 139.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07780668
CAS Number: 29632-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR