2-Bromo-4-iodotoluene 98%

Supplier: Ambeed
Warning

A212182-25G A212182-5G A212182-10G A212182-1G
103818-172EA 6 USD
103818-172 103848-872 77087-320 77087-322 77261-086 77303-234
2-Bromo-4-iodotoluene 98%
2-Bromo-4-iodotoluene

Precaución: for Research Use Only

Formula: C₇H₆BrI
Boiling Pt: 263.6±28.0 °C (760 mm Hg)
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07779014
CAS Number: 26670-89-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR