2-Bromo-4-iodoanisole 95%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-4-iodo-1-methoxybenzene

A102249-100MG A102249-10G A102249-250MG A102249-1G A102249-5G A102249-25G
103796-344EA 16 USD
103796-344 103822-036 76780-536 76780-538 76780-540 77308-912 77308-914 77871-372
2-Bromo-4-iodoanisole 95%
2-Bromo-4-iodoanisole

Precaución: for Research Use Only

Formula: C₇H₆BrIO
Storage Temperature: Refrigerator
MDL Number: MFCD03425935
CAS Number: 182056-39-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR