2-Bromo-4-hydroxybenzaldehyde 95%

Supplier: Ambeed
Warning

A114666-1G A114666-10G A114666-25G
103798-446EA 14 USD
103798-446 103824-288 77311-106 77549-252 77670-606
2-Bromo-4-hydroxybenzaldehyde 95%
2-Bromo-4-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₅BrO₂
MW: 201.02 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16999767
CAS Number: 22532-60-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR