ω-Bromo-4-hydroxyacetophenone 98%
Supplier: Ambeed
|
Danger
|
A172579-25MG
A172579-25G
A172579-5G
103809-056EA
382
USD
103809-056
103845-016
76690-216
77082-212
77082-214
ω-Bromo-4-hydroxyacetophenone 98%
ω-Bromo-4-hydroxyacetophenone
Precaución: for Research Use Only
Formula:
C₈H₇BrO₂ Boiling Pt: 338.7 °C (760 mm Hg) Melting Pt: 124…126 °C Flash Pt: 158.6 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00072424 CAS Number: 2491-38-5 UN: 3261 ADR: 8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...