2-Bromo-4-hydroxy-5-methoxybenzaldehyde 98%

Supplier: Ambeed
Warning

A162285-100G A162285-100MG A162285-10G A162285-250MG A162285-1G A162285-25G A162285-5G
103805-598EA 8 USD
103805-598 103843-848 77235-896 77235-898 77235-900 77235-902 77319-880 77319-882 77319-884
2-Bromo-4-hydroxy-5-methoxybenzaldehyde 98%
2-Bromo-4-hydroxy-5-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇BrO₃
MW: 231.0434 g/mol
Boiling Pt: 326.2 °C (760 mm Hg)
Flash Pt: 151.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01119128
CAS Number: 60632-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR