2-Bromo-4-fluoropyridine 98%

Supplier: Ambeed
Warning

A159960-100G A159960-1G A159960-10G
103805-306EA 14 USD
103805-306 103843-534 76973-510 77319-440 77600-268
2-Bromo-4-fluoropyridine 98%
2-Bromo-4-fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₃BrFN
Boiling Pt: 188.791 °C (760 mm Hg)
Flash Pt: 67.979 °C
Storage Temperature: Ambient
MDL Number: MFCD09260909
CAS Number: 357927-50-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR