α,2-Dibromo-4-fluorotoluene 98%

Supplier: Ambeed
Danger

Synonyms: 2-Bromo-4-fluorobenzylbromide, 2-Bromo-4-fluorobenzyl bromide

A408281-1G A408281-25G A408281-5G A408281-100G A408281-10G
103827-180EA 7 USD
103827-180 103838-226 76841-732 76841-734 76841-736 77143-012 77143-014
α,2-Dibromo-4-fluorotoluene 98%
α,2-Dibromo-4-fluorotoluene

Precaución: for Research Use Only

Formula: C₇H₅Br₂F
Boiling Pt: 254 °C (760 mm Hg)
Flash Pt: 107.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03840524
CAS Number: 61150-57-0
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR