2-Bromo-α,α,α,4-tetrafluorotoluene 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Bromo-4-fluorobenzotrifluoride, 1-Bromo-3-fluoro-6-(trifluoromethyl)benzene
A267091-100G
A267091-10G
A267091-500G
A267091-1G
A267091-25G
A267091-5G
103812-834EA
9
USD
103812-834
103852-736
76772-044
76772-046
76772-048
77092-456
77092-458
77092-460
2-Bromo-α,α,α,4-tetrafluorotoluene 98%
2-Bromo-α,α,α,4-tetrafluorotoluene
Precaución: for Research Use Only
Formula:
C₇H₃BrF₄ MW: 243 g/mol Boiling Pt: 162.1 °C (760 mm Hg) Flash Pt: 78.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD03094041 CAS Number: 351003-21-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...