2-Bromo-4-fluorobenzoic acid 97%
Supplier: Ambeed
|
Warning
|
A205155-25G
A205155-100G
A205155-10G
A205155-500G
A205155-5G
103848-270EA
8
USD
103848-270
77086-472
77260-898
77260-900
77260-902
77302-942
2-Bromo-4-fluorobenzoic acid 97%
2-Bromo-4-fluorobenzoic acid
Precaución: for Research Use Only
Formula:
C₇H₄BrFO₂ Boiling Pt: 294.3 °C (760 mm Hg) Melting Pt: 172…176 °C Flash Pt: 114.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD00055370 CAS Number: 1006-41-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...