2-Bromo-4-fluoroaniline 98%

Supplier: Ambeed
Warning

A143781-100G A143781-10G A143781-500G A143781-5G A143781-25G
103808-644EA 266 USD
103808-644 76972-722 76972-724 76972-726 77299-358 77316-612
2-Bromo-4-fluoroaniline 98%
2-Bromo-4-fluoroaniline

Precaución: for Research Use Only

Formula: C₆H₅BrFN
MW: 190.01 g/mol
Boiling Pt: 221 °C (760 mm Hg)
Melting Pt: 41 °C
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD00042462
CAS Number: 1003-98-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR