2-Bromo-4-fluoro-6-nitroaniline 97%

Supplier: Ambeed
Warning

A184551-1G A184551-25G A184551-5G A184551-10G A184551-250MG
103815-756EA 12 USD
103815-756 103846-320 77083-934 77083-936 77083-938 77096-964 77096-966
2-Bromo-4-fluoro-6-nitroaniline 97%
2-Bromo-4-fluoro-6-nitroaniline

Precaución: for Research Use Only

Formula: C₆H₄BrFN₂O₂
MW: 235.01 g/mol
Boiling Pt: 313.9 °C (760 mm Hg)
Melting Pt: 70…72 °C
Flash Pt: 143.6 °C
Storage Temperature: Ambient
MDL Number: MFCD07779528
CAS Number: 10472-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR