2-Bromo-4-fluoro-6-methylbenzoic acid 97%

Supplier: Ambeed
Warning

A134853-100MG A134853-1MG A134853-250MG
103802-056EA 98 USD
103802-056 77027-264 77027-266 77027-268
2-Bromo-4-fluoro-6-methylbenzoic acid 97%
2-Bromo-4-fluoro-6-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆BrFO₂
MW: 233.03 g/mol
Boiling Pt: 306.6±42.0 °C (760 mm Hg)
Flash Pt: 139.2 °C
Storage Temperature: Ambient
MDL Number: MFCD09263435
CAS Number: 1003709-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR