1-Bromo-5-fluoro-2-nitrobenzene 97%

Supplier: Ambeed
Warning

A156146-10G A156146-500G A156146-1G A156146-25G A156146-5G
103804-832EA 5 USD
103804-832 103843-036 77110-494 77110-496 77318-780 77318-782 77318-784
1-Bromo-5-fluoro-2-nitrobenzene 97%
1-Bromo-5-fluoro-2-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrFNO₂
Melting Pt: 44 °C
Flash Pt: 89.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00792441
CAS Number: 700-36-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR