2-Bromo-4-fluoroanisole 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Bromo-4-fluoro-1-methoxybenzene
A111871-100G
A111871-10G
A111871-25G
A111871-500G
A111871-5G
103823-738EA
5
USD
103823-738
76765-880
76765-882
76765-884
76765-886
77310-548
2-Bromo-4-fluoroanisole 98%
2-Bromo-4-fluoroanisole
Precaución: for Research Use Only
Formula:
C₇H₆BrFO MW: 205.03 g/mol Boiling Pt: 273.6 °C (760 mm Hg) Flash Pt: 98 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012014 CAS Number: 452-08-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...