2-Bromo-4-chloropyridine 97%

Supplier: Ambeed
Warning

A303450-100G A303450-10G A303450-1G A303450-25G A303450-5G A303450-500G
103814-372EA 5 USD
103814-372 103835-540 77002-560 77002-562 77305-868 77305-870 77305-872 77601-366
2-Bromo-4-chloropyridine 97%
2-Bromo-4-chloropyridine

Precaución: for Research Use Only

Formula: C₅H₃BrClN
Boiling Pt: 217.9 °C (760 mm Hg)
Melting Pt: 12…17 °C
Flash Pt: 110 °C
Storage Temperature: Freezer
MDL Number: MFCD03840757
CAS Number: 22918-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR