2-Bromo-4-chlorobenzaldehyde 95%

Supplier: Ambeed
Warning

A255385-100G A255385-10G A255385-500G A255385-5G A255385-25G
103852-050EA 5 USD
103852-050 76815-808 76815-810 76815-812 77091-510 77304-650
2-Bromo-4-chlorobenzaldehyde 95%
2-Bromo-4-chlorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄BrClO
Boiling Pt: 264.6 °C (760 mm Hg)
Melting Pt: 68…73 °C
Flash Pt: 113.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09056792
CAS Number: 84459-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR