2-Bromo-4-chloro-6-methylphenol 97%

Supplier: Ambeed
Warning

A137098-100MG A137098-250MG A137098-1G A137098-5G
103802-404EA 32 USD
103802-404 77010-174 77010-176 77315-356 77315-358
2-Bromo-4-chloro-6-methylphenol 97%
2-Bromo-4-chloro-6-methylphenol

Precaución: for Research Use Only

Formula: C₇H₆BrClO
Storage Temperature: Ambient
MDL Number: MFCD00029750
CAS Number: 54852-68-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR