2-Bromo-4-chloro-5-methylpyridine 97%

Supplier: Ambeed
Warning

A268869-250MG A268869-5G A268869-100MG
103812-940EA 189 USD
103812-940 103852-844 76875-016 77305-026 77883-739
2-Bromo-4-chloro-5-methylpyridine 97%
2-Bromo-4-chloro-5-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅BrClN
MW: 206.47 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11100651
CAS Number: 1033203-40-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR