1-Bromo-5-chloro-2-iodobenzene 98%

Supplier: Ambeed
Warning

A264382-25G A264382-5G A264382-1000G A264382-100G A264382-10G A264382-500G A264382-1G
103812-670EA 5 USD
103812-670 103852-562 77092-220 77092-222 77262-308 77262-310 77262-312 77262-314 77304-892
1-Bromo-5-chloro-2-iodobenzene 98%
1-Bromo-5-chloro-2-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrClI
MW: 317.35 g/mol
Boiling Pt: 278.9 °C (760 mm Hg)
Flash Pt: 122.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00672944
CAS Number: 31928-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR