2'-Bromo-4-chlorobiphenyl 97%

Supplier: Ambeed
Warning

A137697-100G A137697-10G A137697-1G A137697-25G A137697-5G
103802-494EA 5 USD
103802-494 77027-362 77027-364 77315-474 77315-476 77315-478
2'-Bromo-4-chlorobiphenyl 97%
2'-Bromo-4-chlorobiphenyl

Precaución: for Research Use Only

Formula: C₁₂H₈BrCl
Boiling Pt: 325.5 °C (760 mm Hg)
Flash Pt: 173.5 °C
Storage Temperature: Ambient
MDL Number: MFCD18072809
CAS Number: 179526-95-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR