2-Bromo-4,5-dimethylthiazole 97%

Supplier: Ambeed
Warning

A165397-100MG A165397-10G A165397-250MG A165397-5G A165397-1G A165397-25G
103805-964EA 13 USD
103805-964 77028-804 77028-806 77028-808 77028-810 77320-420 77320-422
2-Bromo-4,5-dimethylthiazole 97%
2-Bromo-4,5-dimethylthiazole

Precaución: for Research Use Only

Formula: C₅H₆BrNS
MW: 192.0768 g/mol
Boiling Pt: 223.1 °C (760 mm Hg)
Flash Pt: 88.737 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11501629
CAS Number: 29947-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR