2-Bromo-4,5-dimethylaniline 97%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-4,5-dimethyl aniline

A148695-100MG A148695-250MG A148695-25G A148695-5G A148695-1G
103803-958EA 20 USD
103803-958 76984-546 76984-548 76984-550 76984-552 77299-680
2-Bromo-4,5-dimethylaniline 97%
2-Bromo-4,5-dimethylaniline

Precaución: for Research Use Only

Formula: C₈H₁₀BrN
MW: 200.076 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00031416
CAS Number: 22364-29-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR