6-Bromoveratraldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-3,4-dimethoxybenzaldehyde, 2-Bromo-4,5-dimethoxybenzaldehyde

A233575-100G A233575-10G A233575-500G A233575-5G
103850-650EA 5 USD
103850-650 76771-190 76771-192 76771-194 77303-962
6-Bromoveratraldehyde 98%
6-Bromoveratraldehyde

Precaución: for Research Use Only

Formula: C₉H₉BrO₃
MW: 245.07 g/mol
Boiling Pt: 316.1 °C (760 mm Hg)
Melting Pt: 150…151 °C
Flash Pt: 145 °C
Storage Temperature: Ambient
MDL Number: MFCD00003301
CAS Number: 5392-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR