2-Bromo-4,5-difluorobenzoic acid 98%

Supplier: Ambeed
Warning

A114704-100G A114704-10G A114704-25G A114704-500G A114704-1G A114704-5G
103798-454EA 9 USD
103798-454 103824-298 77257-846 77257-848 77257-850 77257-852 77297-320 77311-112
2-Bromo-4,5-difluorobenzoic acid 98%
2-Bromo-4,5-difluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃BrF₂O₂
Melting Pt: 118…122 °C
Flash Pt: 130 °C
Storage Temperature: Ambient
MDL Number: MFCD04973422
CAS Number: 64695-84-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR