2-Bromo-4,5-difluorobenzaldehyde 97%

Supplier: Ambeed
Warning

A126662-100G A126662-10G A126662-250MG A126662-25G A126662-1G A126662-5G
103800-848EA 9 USD
103800-848 76782-068 76782-070 76782-072 77298-142 77313-078 77313-080
2-Bromo-4,5-difluorobenzaldehyde 97%
2-Bromo-4,5-difluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃BrF₂O
Boiling Pt: 232.0±35.0 °C (760 mm Hg)
Flash Pt: 94.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD16659662
CAS Number: 476620-54-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR